accessibility menu, dialog, popup

UserName

CAS RN 161814-49-9

CH3CH3N(R)HO(S)NHOO(S)OSOONH2

CAS RN 161814-49-9

IUPAC Name: (3S)-oxolan-3-yl N-[(2S,3R)-3-hydroxy-4-[N-(2-methylpropyl)4-aminobenzenesulfonamido]-1-phenylbutan-2-yl]carbamate
Synonyms: amprenavir agenerase prozei AMV APV vertex
Molecular Weight (g/mol): 505.63
Molecular Formula: C25H35N3O6S
InChi Key: YMARZQAQMVYCKC-OEMFJLHTSA-N
SMILES: CC(C)CN(C[C@@H](O)[C@H](CC1=CC=CC=C1)NC(=O)O[C@H]1CCOC1)S(=O)(=O)C1=CC=C(N)C=C1
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results