accessibility menu, dialog, popup

UserName

OSO(Z)

CAS RN 16212-06-9

IUPAC Name: [(1Z)-2-(benzenesulfonyl)ethenyl]benzene
Synonyms: [(1Z)-2-(benzenesulfonyl)ethenyl]benzene
Molecular Weight (g/mol): 244.31
Molecular Formula: C14H12O2S
InChi Key: DNMCCXFLTURVLK-QXMHVHEDSA-N
SMILES: O=S(=O)(\C=C/C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results