accessibility menu, dialog, popup

UserName

CAS RN 16219-75-3

H3C(E)

CAS RN 16219-75-3

IUPAC Name: (5E)-5-ethylidenebicyclo[2.2.1]hept-2-ene
Synonyms: ethylidene norbornene
Molecular Weight (g/mol): 120.2
Molecular Formula: C9H12
InChi Key: OJOWICOBYCXEKR-KRXBUXKQNA-N
SMILES: C\C=C1/CC2CC1C=C2
Molecular Weight (g/mol) 
  • (4)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (3)
  • (4)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)