accessibility menu, dialog, popup

UserName

CAS RN 16245-79-7

H3CNH2

CAS RN 16245-79-7

IUPAC Name: 4-octylaniline
Synonyms: 4-octylaniline
Molecular Weight (g/mol): 205.35
Molecular Formula: C14H23N
InChi Key: ORKQJTBYQZITLA-UHFFFAOYSA-N
SMILES: CCCCCCCCC1=CC=C(N)C=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results