accessibility menu, dialog, popup

UserName

CAS RN 162870-29-3

OHHOOHONH2

CAS RN 162870-29-3

IUPAC Name: 2-amino-2-(3,5-dihydroxyphenyl)acetic acid
Synonyms: dihydroxyphenylglycine
Molecular Weight (g/mol): 183.16
Molecular Formula: C8H9NO4
InChi Key: HOOWCUZPEFNHDT-UHFFFAOYNA-N
SMILES: NC(C(O)=O)C1=CC(O)=CC(O)=C1
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)