accessibility menu, dialog, popup

UserName

CAS RN 163042-96-4

H3CNHO(S)O(R)(R)OH(S)OHNNNHINClN

CAS RN 163042-96-4

IUPAC Name: (2S,3S,4R,5R)-5-(2-chloro-6-{[(3-iodophenyl)methyl]amino}-9H-purin-9-yl)-3,4-dihydroxy-N-methyloxolane-2-carboxamide
Synonyms: (2S,3S,4R,5R)-5-(2-chloro-6-{[(3-iodophenyl)methyl]amino}purin-9-yl)-3,4-dihydroxy-N-methyloxolane-2-carboxamide
Molecular Weight (g/mol): 544.73
Molecular Formula: C18H18ClIN6O4
InChi Key: IPSYPUKKXMNCNQ-PFHKOEEOSA-N
SMILES: CNC(=O)[C@H]1O[C@H]([C@H](O)[C@@H]1O)N1C=NC2=C(NCC3=CC=CC(I)=C3)N=C(Cl)N=C12
Grade 
  • (6)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (1)
  • (5)
  • (1)
  • (6)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)