CAS RN 163169-29-7
CAS RN 163169-29-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Promotions Available
(S,S)-[2-(4'-Isopropyloxazolin-2'yl)ferrocenyl]diphenylphosphine, 97%, Thermo Scientific Chemicals
(S,S)-[2-(4'-Isopropyloxazolin-2'yl)ferrocenyl]diphenylphosphine, 97%, Thermo Scientific Chemicals
CAS: 163169-29-7 Molecular Formula: C28H28FeNOP Molecular Weight (g/mol): 481.36 MDL Number: MFCD15144869 InChI Key: FELCHLOAEMNOQM-KISGOYOSNA-N Synonym: (S)[(Sp)-2-(Diphenylphosphino)ferrocenyl]-4-isopropyloxazoline IUPAC Name: (S,S)-[2-(4-Isopropyl-2-oxazolinyl)ferrocenyl]diphenylphosphine SMILES: [Fe].c1cccc1.CC(C)[C@H]1COC(=N1)c1cccc1P(C1=CC=CC=C1)C1=CC=CC=C1
2
(S,S)-[2-(4-Isopropyl-2-oxazolinyl)ferrocenyl]diphenylphosphine, 97%, Thermo Scientific Chemicals
CAS: 163169-29-7 Molecular Formula: C28H28FeNOP Molecular Weight (g/mol): 481.36 MDL Number: MFCD15144869 InChI Key: FELCHLOAEMNOQM-KISGOYOSNA-N IUPAC Name: (S,S)-[2-(4-Isopropyl-2-oxazolinyl)ferrocenyl]diphenylphosphine SMILES: [Fe].c1cccc1.CC(C)[C@H]1COC(=N1)c1cccc1P(C1=CC=CC=C1)C1=CC=CC=C1