accessibility menu, dialog, popup

UserName

CAS RN 16323-43-6

IUPAC Name: (2E)-3-{4-[(1E)-2-carboxyeth-1-en-1-yl]phenyl}prop-2-enoic acid
Synonyms: (2E)-3-{4-[(1E)-2-carboxyeth-1-en-1-yl]phenyl}prop-2-enoic acid
Molecular Weight (g/mol): 218.21
Molecular Formula: C12H10O4
InChi Key: AAFXQFIGKBLKMC-KQQUZDAGSA-N
SMILES: OC(=O)\C=C\C1=CC=C(\C=C\C(O)=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results