accessibility menu, dialog, popup

UserName

CH3OOCH3(Z)(Z)(Z)

CAS RN 16326-32-2

IUPAC Name: methyl (6Z,9Z,12Z)-octadeca-6,9,12-trienoate
Synonyms: methyl (6Z,9Z,12Z)-octadeca-6,9,12-trienoate
Molecular Weight (g/mol): 292.46
Molecular Formula: C19H32O2
InChi Key: JFRWATCOFCPIBM-JPFHKJGASA-N
SMILES: CCCCC\C=C/C\C=C/C\C=C/CCCCC(=O)OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results