accessibility menu, dialog, popup

UserName

CAS RN 163439-82-5

OH(R)NH(R)HO

CAS RN 163439-82-5

IUPAC Name: (3R,4R)-1-benzyl-3,4-dihydroxypyrrolidin-1-ium
Synonyms: (3R,4R)-1-benzyl-3,4-dihydroxypyrrolidin-1-ium
Molecular Weight (g/mol): 194.25
Molecular Formula: C11H16NO2
InChi Key: QJRIUWQPJVPYSO-GHMZBOCLSA-O
SMILES: O[C@@H]1C[NH+](CC2=CC=CC=C2)C[C@H]1O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results