accessibility menu, dialog, popup

UserName

CAS RN 163444-17-5

FFFINH2

CAS RN 163444-17-5

IUPAC Name: 2-iodo-4-(trifluoromethyl)aniline
Synonyms: 2-iodo-4-(trifluoromethyl)aniline
Molecular Weight (g/mol): 287.02
Molecular Formula: C7H5F3IN
InChi Key: UKKWTZPXYIYONW-UHFFFAOYSA-N
SMILES: NC1=C(I)C=C(C=C1)C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results