CAS RN 164033-12-9
IUPAC Name:
(1-isocyanatopropyl)benzene
Synonyms:
(1-isocyanatopropyl)benzene
Molecular Weight (g/mol):
161.2
Molecular Formula:
C10H11NO
InChi Key:
XGPXXSJFZSZULR-UHFFFAOYNA-N
SMILES:
CCC(N=C=O)C1=CC=CC=C1