accessibility menu, dialog, popup

UserName

CAS RN 1641-09-4

NS

CAS RN 1641-09-4

IUPAC Name: thiophene-3-carbonitrile
Synonyms: thiophene-3-carbonitrile
Molecular Weight (g/mol): 109.15
Molecular Formula: C5H3NS
InChi Key: GSXCEVHRIVLFJV-UHFFFAOYSA-N
SMILES: N#CC1=CSC=C1
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)