CAS RN 1642-54-2
CAS RN 1642-54-2
IUPAC Name:
2-hydroxypropane-1,2,3-tricarboxylic acid; N,N-diethyl-4-methylpiperazine-1-carboxamide
Synonyms:
citric acid; diethylcarbamazine
Molecular Weight (g/mol):
391.42
Molecular Formula:
C16H29N3O8
InChi Key:
PGNKBEARDDELNB-UHFFFAOYSA-N
SMILES:
OC(=O)CC(O)(CC(O)=O)C(O)=O.CCN(CC)C(=O)N1CCN(C)CC1
1
–
3
of
3
results
1
Diethylcarbamazine Citrate, LoGiCal
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
2
Diethylcarbamazine Citrate, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3
Diethylcarbamazine citrate, MedChemExpress
MedChemExpress Diethylcarbamazine citrate is an inhibitor of arachidonic acid metabolism in filarial microfilaria; is highly specific for several parasites and does not contain any toxic metallic elements.