accessibility menu, dialog, popup

UserName

H2CCH2

CAS RN 1647-16-1

IUPAC Name: deca-1,9-diene
Synonyms: 1,9-decadiene
Molecular Weight (g/mol): 138.25
Molecular Formula: C10H18
InChi Key: NLDGJRWPPOSWLC-UHFFFAOYSA-N
SMILES: C=CCCCCCCC=C
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)