accessibility menu, dialog, popup

UserName

CAS RN 165904-20-1

BOOH3CCH3H3CH3CSiCH3H3CCH3

CAS RN 165904-20-1

IUPAC Name: trimethyl[2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)ethyl]silane
Synonyms: trimethyl[2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)ethyl]silane
Molecular Weight (g/mol): 228.21
Molecular Formula: C11H25BO2Si
InChi Key: YKYAZAJUMAWTGO-UHFFFAOYSA-N
SMILES: CC1(C)OB(CC[Si](C)(C)C)OC1(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results