CAS RN 16593-81-0
IUPAC Name:
(4E)-3-hydroxy-4-[2-(pyridin-2-yl)hydrazin-1-ylidene]cyclohexa-2,5-dien-1-one
Synonyms:
(4E)-3-hydroxy-4-[2-(pyridin-2-yl)hydrazin-1-ylidene]cyclohexa-2,5-dien-1-one
Molecular Weight (g/mol):
215.21
Molecular Formula:
C11H9N3O2
InChi Key:
VLCAILLZPUINNF-UKTHLTGXSA-N
SMILES:
OC1=CC(=O)C=C\C1=N/NC1=CC=CC=N1
1
–
2
of
2
results
1
Promotions Available
Thermo Scientific Chemicals 4-(2-Pyridylazo)resorcinol monosodium salt monohydrate, 99+%
Thermo Scientific Chemicals 4-(2-Pyridylazo)resorcinol monosodium salt monohydrate, 99+%
CAS: 16593-81-0 | C11H9N3O2 | 215.21 g/mol
2
4-(2-Pyridylazo)resorcinol monosodium salt monohydrate
CAS: 16593-81-0 Molecular Formula: C11H9N3O2 Molecular Weight (g/mol): 215.21 MDL Number: MFCD00150712,MFCD00150712,MFCD00150712,MFCD00006256 InChI Key: VLCAILLZPUINNF-UKTHLTGXSA-N Synonym: 4-2-pyridylazo resorcinol monosodium saltmonohydrate PubChem CID: 132988475 IUPAC Name: 3-hydroxy-4-(pyridin-2-ylhydrazinylidene)cyclohexa-2,5-dien-1-one;sodium;hydrate SMILES: OC1=CC(=O)C=C\C1=N/NC1=CC=CC=N1