accessibility menu, dialog, popup

UserName

SOOHHNOO

CAS RN 167015-11-4

IUPAC Name: 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-[(triphenylmethyl)sulfanyl]propanoic acid
Synonyms: 2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-[(triphenylmethyl)sulfanyl]propanoic acid
Molecular Weight (g/mol): 585.72
Molecular Formula: C37H31NO4S
InChi Key: KLBPUVPNPAJWHZ-UHFFFAOYNA-N
SMILES: OC(=O)C(CSC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results