accessibility menu, dialog, popup

UserName

CAS RN 167834-24-4

H3COO(S)NH3

CAS RN 167834-24-4

IUPAC Name: (1S)-3-ethoxy-3-oxo-1-phenylpropan-1-aminium
Synonyms: (1S)-3-ethoxy-3-oxo-1-phenylpropan-1-aminium
Molecular Weight (g/mol): 194.25
Molecular Formula: C11H16NO2
InChi Key: NUWRDXMXYDWUAN-JTQLQIEISA-O
SMILES: CCOC(=O)C[C@H]([NH3+])C1=CC=CC=C1
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)