accessibility menu, dialog, popup

UserName

CAS RN 16793-91-2

BrCl

CAS RN 16793-91-2

IUPAC Name: 1-(2-bromoethyl)-2-chlorobenzene
Synonyms: 1-(2-bromoethyl)-2-chlorobenzene
Molecular Weight (g/mol): 219.51
Molecular Formula: C8H8BrCl
InChi Key: AECBVDLERUETKG-UHFFFAOYSA-N
SMILES: ClC1=CC=CC=C1CCBr

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results