accessibility menu, dialog, popup

UserName

CAS RN 1681-37-4

NH2NHNOO

CAS RN 1681-37-4

IUPAC Name: 4-amino-3-nitropyridin-1-ium
Synonyms: 4-amino-3-nitropyridin-1-ium
Molecular Weight (g/mol): 140.12
Molecular Formula: C5H6N3O2
InChi Key: IUPPEELMBOPLDJ-UHFFFAOYSA-O
SMILES: NC1=CC=[NH+]C=C1[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results