accessibility menu, dialog, popup

UserName

CH3NH3CH3CH3COHSOOO

CAS RN 16873-13-5

IUPAC Name: tetraethylazanium hydrogen sulfate
Synonyms: tetraethylammonium hydrogen sulfate
Molecular Weight (g/mol): 227.32
Molecular Formula: C8H21NO4S
InChi Key: CREVBWLEPKAZBH-UHFFFAOYSA-M
SMILES: OS([O-])(=O)=O.CC[N+](CC)(CC)CC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results