accessibility menu, dialog, popup

UserName

OHIIN

CAS RN 1689-83-4

IUPAC Name: 4-hydroxy-3,5-diiodobenzonitrile
Synonyms: ioxynil
Molecular Weight (g/mol): 370.92
Molecular Formula: C7H3I2NO
InChi Key: NRXQIUSYPAHGNM-UHFFFAOYSA-N
SMILES: OC1=C(I)C=C(C=C1I)C#N
Quantity 
  • (1)
  • (1)
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results