accessibility menu, dialog, popup

UserName

CAS RN 16948-16-6

CH3(S)OOHNCH3OOH3CH3CCH3

CAS RN 16948-16-6

IUPAC Name: 2-{[(tert-butoxy)carbonyl](methyl)amino}propanoic acid
Synonyms: 2-[(tert-butoxycarbonyl)(methyl)amino]propanoic acid
Molecular Weight (g/mol): 203.24
Molecular Formula: C9H17NO4
InChi Key: VLHQXRIIQSTJCQ-UHFFFAOYNA-N
SMILES: CC(N(C)C(=O)OC(C)(C)C)C(O)=O
Quantity 
  • (3)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)