accessibility menu, dialog, popup

UserName

CAS RN 1701-20-8

H3CHNOFFF

CAS RN 1701-20-8

IUPAC Name: 6-methyl-2-(trifluoromethyl)-1,4-dihydroquinolin-4-one
Synonyms: 6-methyl-2-(trifluoromethyl)-1H-quinolin-4-one
Molecular Weight (g/mol): 227.19
Molecular Formula: C11H8F3NO
InChi Key: UNVMZLUVACVTDT-UHFFFAOYSA-N
SMILES: CC1=CC=C2NC(=CC(=O)C2=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results