accessibility menu, dialog, popup

UserName

CAS RN 1710-98-1

CH3H3CCH3ClO

CAS RN 1710-98-1

IUPAC Name: 4-tert-butylbenzoyl chloride
Synonyms: 4-tert-butylbenzoyl chloride
Molecular Weight (g/mol): 196.67
Molecular Formula: C11H13ClO
InChi Key: WNLMYNASWOULQY-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC=C(C=C1)C(Cl)=O
Boiling Point 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)