CAS RN 171179-06-9
CAS RN 171179-06-9
IUPAC Name:
N4-(3-bromophenyl)-N6-methylpyrido[3,4-d]pyrimidine-4,6-diamine
Synonyms:
N4-(3-bromophenyl)-N6-methylpyrido[3,4-d]pyrimidine-4,6-diamine
Molecular Weight (g/mol):
330.19
Molecular Formula:
C14H12BrN5
InChi Key:
KFHMLBXBRCITHF-UHFFFAOYSA-N
SMILES:
CNC1=NC=C2N=CN=C(NC3=CC(Br)=CC=C3)C2=C1
1
–
2
of
2
results
1
Tocris Bioscience™ PD 158780
CAS: 171179-06-9 Molecular Formula: C14H12BrN5
2
PD158780, MedChemExpress
MedChemExpress PD158780 is a potent EGFR family inhibitor with IC50s of 8 pM, 49, 52, 52 nM for EGFR, ErbB2, ErbB3, and ErbB4, respectively.