accessibility menu, dialog, popup

UserName

CH3(S)(S)H3COHNNNONNO(R)O(R)NNNFF

CAS RN 171228-49-2

IUPAC Name: 4-{4-[4-(4-{[(3R,5R)-5-(2,4-difluorophenyl)-5-[(1H-1,2,4-triazol-1-yl)methyl]oxolan-3-yl]methoxy}phenyl)piperazin-1-yl]phenyl}-1-[(2S,3S)-2-hydroxypentan-3-yl]-4,5-dihydro-1H-1,2,4-triazol-5-one
Synonyms: posaconazole
Molecular Weight (g/mol): 700.79
Molecular Formula: C37H42F2N8O4
InChi Key: RAGOYPUPXAKGKH-XAKZXMRKSA-N
SMILES: CC[C@@H]([C@H](C)O)N1N=CN(C1=O)C1=CC=C(C=C1)N1CCN(CC1)C1=CC=C(OC[C@@H]2CO[C@](CN3C=NC=N3)(C2)C2=CC=C(F)C=C2F)C=C1
Percent Purity 
  • (3)
  • (4)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
Grade 
  • (1)
  • (4)
Color 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results