accessibility menu, dialog, popup

UserName

CAS RN 171364-82-2

CH3H3COBOH3CH3CN

CAS RN 171364-82-2

IUPAC Name: 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
Synonyms: 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
Molecular Weight (g/mol): 229.09
Molecular Formula: C13H16BNO2
InChi Key: HOPDTPGXBZCBNP-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=C(C=C1)C#N
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)