accessibility menu, dialog, popup

UserName

CAS RN 17289-20-2

CH3OONNH3C

CAS RN 17289-20-2

IUPAC Name: methyl 1-methyl-1H-imidazole-5-carboxylate
Synonyms: methyl 3-methylimidazole-4-carboxylate
Molecular Weight (g/mol): 140.14
Molecular Formula: C6H8N2O2
InChi Key: AKDPLDCXQNEMCL-UHFFFAOYSA-N
SMILES: COC(=O)C1=CN=CN1C
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)