accessibility menu, dialog, popup

UserName

CAS RN 172975-69-8

CH3CH3BOHHO

CAS RN 172975-69-8

IUPAC Name: (3,5-dimethylphenyl)boronic acid
Synonyms: 3,5-dimethylphenylboronic acid
Molecular Weight (g/mol): 149.98
Molecular Formula: C8H11BO2
InChi Key: DJGHSJBYKIQHIK-UHFFFAOYSA-N
SMILES: CC1=CC(=CC(C)=C1)B(O)O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)