accessibility menu, dialog, popup

UserName

CAS RN 1730-25-2

H2CMgBr

CAS RN 1730-25-2

IUPAC Name: allylmagnesium bromide
Synonyms: allylmagnesium bromide
Molecular Weight (g/mol): 145.28
Molecular Formula: C3H5BrMg
InChi Key: DQEUYIQDSMINEY-UHFFFAOYSA-M
SMILES: [Mg++].[Br-].[CH2-]C=C
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results