accessibility menu, dialog, popup

UserName

H3COOHNH2

CAS RN 17350-84-4

IUPAC Name: 2-amino-2-methyl-3-phenylpropanoic acid
Synonyms: 2-amino-2-methyl-3-phenylpropanoic acid
Molecular Weight (g/mol): 179.22
Molecular Formula: C10H13NO2
InChi Key: HYOWVAAEQCNGLE-UHFFFAOYNA-N
SMILES: CC(N)(CC1=CC=CC=C1)C(O)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results