CAS RN 173603-23-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(E)-alpha,beta-Styrenediboronic acid bis(pinacol) ester, 98%, Thermo Scientific Chemicals
CAS: 173603-23-1 Molecular Formula: C20H30B2O4 MDL Number: MFCD02683500 Synonym: cis-1,2-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)styrene