CAS RN 17374-26-4
CAS RN 17374-26-4
IUPAC Name:
4,5,6,7-tetrabromo-2H-1,2,3-benzotriazole
Synonyms:
4,5,6,7-tetrabromo-2H-1,2,3-benzotriazole
Molecular Weight (g/mol):
434.71
Molecular Formula:
C6HBr4N3
InChi Key:
OMZYUVOATZSGJY-UHFFFAOYSA-N
SMILES:
BrC1=C(Br)C(Br)=C(Br)C2=NNN=C12
1
–
2
of
2
results
1
Tocris Bioscience™ TBB
CAS: 17374-26-4 Molecular Formula: C6HBr4N3
2
TBB, MedChemExpress
MedChemExpress TBB is a cell-permeable and ATP-competitive CK2 inhibitor with an IC50 of 0.15 μM for rat liver CK2.