accessibility menu, dialog, popup

UserName

CH3H3CCH3OONNO

CAS RN 174855-57-3

IUPAC Name: tert-butyl 4-(2-formylphenyl)piperazine-1-carboxylate
Synonyms: tert-butyl 4-(2-formylphenyl)piperazine-1-carboxylate
Molecular Weight (g/mol): 290.36
Molecular Formula: C16H22N2O3
InChi Key: FGJACYJASSSXNJ-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCN(CC1)C1=CC=CC=C1C=O
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results