CAS RN 175026-96-7
CAS RN 175026-96-7
IUPAC Name:
ethyl 8,15-dioxo-14-azatetracyclo[7.7.1.0²,⁷.0¹³,¹⁷]heptadeca-1(16),2,4,6,9(17),10,12-heptaene-16-carboxylate
Synonyms:
ethyl 8,15-dioxo-14-azatetracyclo[7.7.1.0²,⁷.0¹³,¹⁷]heptadeca-1(16),2,4,6,9(17),10,12-heptaene-16-carboxylate
Molecular Weight (g/mol):
319.32
Molecular Formula:
C19H13NO4
InChi Key:
UFJGFNHRMPMALC-UHFFFAOYSA-N
SMILES:
CCOC(=O)C1=C2C3=CC=CC=C3C(=O)C3=C2C(NC1=O)=CC=C3
1
–
1
of
1
results
1
NQDI-1, MedChemExpress
MedChemExpress NQDI-1 inhibits apoptosis signal-regulating kinase 1 (ASK1) with a Ki of 500 nM and an IC50 of 3 μM.