CAS RN 175140-00-8
CAS RN 175140-00-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
CP 376395, MedChemExpress
MedChemExpress CP 376395 is a potent and selective Corticotropin releasing factor 1 (CRF1) receptor antagonist.