CAS RN 1752-96-1
CAS RN 1752-96-1
IUPAC Name:
N-phenylpyridine-3-carboxamide
Synonyms:
nicotinanilide
Molecular Weight (g/mol):
198.23
Molecular Formula:
C12H10N2O
InChi Key:
NYQXIOZBHWFCBU-UHFFFAOYSA-N
SMILES:
O=C(NC1=CC=CC=C1)C1=CC=CN=C1
1
–
3
of
3
results
1
Nicotinic Anilide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Nicotinic Anilide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Nicotinic Anilide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.