accessibility menu, dialog, popup

UserName

CAS RN 175204-90-7

NOHOOFFF

CAS RN 175204-90-7

IUPAC Name: 6-(2,2,2-trifluoroethoxy)pyridine-3-carboxylic acid
Synonyms: 6-(2,2,2-trifluoroethoxy)pyridine-3-carboxylic acid
Molecular Weight (g/mol): 221.14
Molecular Formula: C8H6F3NO3
InChi Key: GZOOLXWQKURRPU-UHFFFAOYSA-N
SMILES: OC(=O)C1=CN=C(OCC(F)(F)F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results