accessibility menu, dialog, popup

UserName

CH3OOONH2

CAS RN 175205-10-4

IUPAC Name: 7-amino-4-(methoxymethyl)-2H-chromen-2-one
Synonyms: 7-amino-4-(methoxymethyl)chromen-2-one
Molecular Weight (g/mol): 205.21
Molecular Formula: C11H11NO3
InChi Key: QZZLLHOMMCKWIQ-UHFFFAOYSA-N
SMILES: COCC1=CC(=O)OC2=CC(N)=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results