CAS RN 175276-52-5
CAS RN 175276-52-5
IUPAC Name:
ethyl 4-formyl-2,5-dimethyl-1-phenyl-1H-pyrrole-3-carboxylate
Synonyms:
ethyl 4-formyl-2,5-dimethyl-1-phenylpyrrole-3-carboxylate
Molecular Weight (g/mol):
271.32
Molecular Formula:
C16H17NO3
InChi Key:
BGDYIZGTPVRWAM-UHFFFAOYSA-N
SMILES:
CCOC(=O)C1=C(C)N(C(C)=C1C=O)C1=CC=CC=C1
1
–
1
of
1
results
1
ethyle4-formyl-2,5-dimethyl-1-phenyl-1H-pyrrole-3-carboxylate, 97%, Thermo Scientific™
CAS: 175276-52-5 Molecular Formula: C16H17NO3 Molecular Weight (g/mol): 271.316 MDL Number: MFCD00204225 InChI Key: BGDYIZGTPVRWAM-UHFFFAOYSA-N Synonym: ethyl 4-formyl-2,5-dimethyl-1-phenyl-1h-pyrrole-3-carboxylate,4-formyl-2,5-dimethyl-1-phenyl-1h-pyrrole-3-carboxylic acid ethyl ester PubChem CID: 2798484 IUPAC Name: ethyl 4-formyl-2,5-dimethyl-1-phenylpyrrole-3-carboxylate SMILES: CCOC(=O)C1=C(N(C(=C1C=O)C)C2=CC=CC=C2)C