accessibility menu, dialog, popup

UserName

BOOH3CCH3H3CH3CSBOOCH3H3CCH3CH3

CAS RN 175361-81-6

IUPAC Name: 4,4,5,5-tetramethyl-2-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophen-2-yl]-1,3,2-dioxaborolane
Synonyms: 4,4,5,5-tetramethyl-2-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophen-2-yl]-1,3,2-dioxaborolane
Molecular Weight (g/mol): 336.06
Molecular Formula: C16H26B2O4S
InChi Key: AOJXAKMKFDBHHT-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=C(S1)B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results