accessibility menu, dialog, popup

UserName

CAS RN 176548-70-2

OOFBr

CAS RN 176548-70-2

IUPAC Name: 3-bromo-5-fluorobenzoate
Synonyms: 3-bromo-5-fluorobenzoate
Molecular Weight (g/mol): 218
Molecular Formula: C7H3BrFO2
InChi Key: KLSLJMGWUPAQGZ-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC(F)=CC(Br)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results