accessibility menu, dialog, popup

UserName

BOOH3COOCH3HOOH

CAS RN 177735-55-6

IUPAC Name: [3,5-bis(methoxycarbonyl)phenyl]boronic acid
Synonyms: 3,5-bis(methoxycarbonyl)phenylboronic acid
Molecular Weight (g/mol): 238
Molecular Formula: C10H11BO6
InChi Key: WEJWFDLAZSVCJK-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(=CC(=C1)B(O)O)C(=O)OC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results