accessibility menu, dialog, popup

UserName

FFFNFFF

CAS RN 177952-38-4

IUPAC Name: 2,4-bis(trifluoromethyl)benzonitrile
Synonyms: 2,4-bis(trifluoromethyl)benzonitrile
Molecular Weight (g/mol): 239.12
Molecular Formula: C9H3F6N
InChi Key: RQKXUWGXSNMDDS-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=C(C#N)C(=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results