accessibility menu, dialog, popup

UserName

CAS RN 1784-03-8

HClH3COONH2NNH2NHSOOCH3

CAS RN 1784-03-8

IUPAC Name: hydrogen methyl 5-[(diaminomethylidene)amino]-2-(4-methylbenzenesulfonamido)pentanoate chloride
Synonyms: hydrogen methyl 5-[(diaminomethylidene)amino]-2-(4-methylbenzenesulfonamido)pentanoate chloride
Molecular Weight (g/mol): 378.87
Molecular Formula: C14H23ClN4O4S
InChi Key: JIQFFACVQXXHMY-UHFFFAOYNA-N
SMILES: [H+].[Cl-].COC(=O)C(CCCN=C(N)N)NS(=O)(=O)C1=CC=C(C)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (3)