accessibility menu, dialog, popup

UserName

H3CNNCH3I

CAS RN 178631-05-5

IUPAC Name: 1-hexyl-3-methyl-1H-imidazol-3-ium iodide
Synonyms: 1-hexyl-3-methylimidazolium iodide
Molecular Weight (g/mol): 294.18
Molecular Formula: C10H19IN2
InChi Key: CZIUVCSYOGFUPH-UHFFFAOYSA-M
SMILES: [I-].CCCCCCN1C=C[N+](C)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results