CAS RN 178946-89-9
CAS RN 178946-89-9
IUPAC Name:
3-[(4-chlorophenyl)(1H-indol-3-yl)methyl]-1H-indole
Synonyms:
3-[(4-chlorophenyl)(1H-indol-3-yl)methyl]-1H-indole
Molecular Weight (g/mol):
356.85
Molecular Formula:
C23H17ClN2
InChi Key:
LTLRXTDMXOFBDW-UHFFFAOYSA-N
SMILES:
ClC1=CC=C(C=C1)C(C1=CNC2=CC=CC=C12)C1=CNC2=CC=CC=C12
1
–
2
of
2
results
1
Tocris Bioscience™ C-DIM 12
CAS: 178946-89-9 Molecular Formula: C23H17ClN2
2
C-DIM12, MedChemExpress
MedChemExpress C-DIM12 is a synthetic Nurr1 activaor induces Nurr1 and DA gene expression in cell lines and primary neurons.