accessibility menu, dialog, popup

UserName

CAS RN 17920-35-3

H3CONH2N

CAS RN 17920-35-3

IUPAC Name: 6-methoxypyridin-2-amine
Synonyms: 6-methoxypyridin-2-amine
Molecular Weight (g/mol): 124.14
Molecular Formula: C6H8N2O
InChi Key: DEUALFRBMNMGDS-UHFFFAOYSA-N
SMILES: COC1=CC=CC(N)=N1
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)